0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
2,3'-Bipyridine
2,3'-Bipyridine 95%
2,3'-Bipyridine 95%
Bottle, 1 g
APOSOR931059-1G
:
Apollo Scientific
2,3'-Bipyridine 95%
APOSOR931059-1G
:
Apollo Scientific
:
581-50-0
2,3'-Bipyridine 95%
Bottle, 1 g
:
581-50-0
21,80 €
1 * 1 g
-
+
:
581-50-0
:
C₁₀H₈N₂
:
MFCD00006215
:
156.184 g/mol
:
Ambient
Pack type:
Bottle
Pk:
1 g