Order Entry
Export
Orders LinkContactUsLinkComponent
1,5-Naphthyridine

1,5-Naphthyridine

Bottle, 1 g
Catalog # APOSOR40671-1G
Corrosive
Irritant
Supplier:   Apollo Scientific
1,5-Naphthyridine
Catalog # APOSOR40671-1G
Supplier:  Apollo Scientific
CAS Number:  254-79-5
1,5-Naphthyridine

Bottle, 1 g

CAS Number:  254-79-5
Price
21,80 €
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:254-79-5
  • Formula:C₈H₆N₂
  • MDL Number:MFCD00039727
  • Molecular Weight:130.15 g/mol
  • Boiling Pt:150…152 °C/54mmHg
  • Melting Pt:69…75 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications