Order Entry
Export
Orders LinkContactUsLinkComponent
 

1,9-Nonanedithiol 95+%

Bottle, 25 g
Catalog # APOSOR451302-25G
Irritant
Supplier:   Apollo Scientific
1,9-Nonanedithiol 95+%
Catalog # APOSOR451302-25G
Supplier:  Apollo Scientific
CAS Number:  3489-28-9
1,9-Nonanedithiol 95+%

Bottle, 25 g

CAS Number:  3489-28-9
Price
137,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  1,9-Dimercaptononane
  • CAS Number:3489-28-9
  • Formula:C₉H₂₀S₂
  • MDL Number:MFCD00004914
  • Molecular Weight:192.39 g/mol
  • Boiling Pt:284 °C
  • Density:0.952 g/cm³ (25 °C)
  • Flash Pt:102 °C
  • Storage Temperature:Ambient
  • UN:3334
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications