1,9-Nonanedithiol 95+%
Bottle, 1 g
Catalog # APOSOR451302-1G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: 1,9-Dimercaptononane
- CAS Number:3489-28-9
- Formula:C₉H₂₀S₂
- MDL Number:MFCD00004914
- Molecular Weight:192.39 g/mol
- Boiling Pt:284 °C
- Density:0.952 g/cm³ (25 °C)
- Flash Pt:102 °C
- Storage Temperature:Ambient
- UN:3334
- ADR:9,III
Specifications
Specifications
- Pack type:Bottle
- Pk:1 g
- Cat. No.:APOSOR451302-1G



