Order Entry
Export
Orders LinkContactUsLinkComponent
 

2,6-Diethylpyridine 95%

Bottle, 1 g
Catalog # APOSOR928697-1G
Supplier:   Apollo Scientific
2,6-Diethylpyridine 95%
Catalog # APOSOR928697-1G
Supplier:  Apollo Scientific
CAS Number:  935-28-4
2,6-Diethylpyridine 95%

Bottle, 1 g

CAS Number:  935-28-4
Price
34,80 €
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:935-28-4
  • Formula:C₉H₁₃N
  • MDL Number:MFCD00049215
  • Molecular Weight:135.206 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications