Order Entry
Export
Orders LinkContactUsLinkComponent
 

3,3-Diphenylpropylamine 98%

Bottle, 5 g
Catalog # APOSOR5085-5G
Irritant
Supplier:   Apollo Scientific
3,3-Diphenylpropylamine 98%
Catalog # APOSOR5085-5G
Supplier:  Apollo Scientific
CAS Number:  5586-73-2
3,3-Diphenylpropylamine 98%

Bottle, 5 g

CAS Number:  5586-73-2
Price
36,20 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  3,3-Diphenylpropan-1-amine
  • CAS Number:5586-73-2
  • Formula:C₁₅H₁₇N
  • MDL Number:MFCD00008202
  • Molecular Weight:211.31 g/mol
  • Boiling Pt:166 °C/2mmHg
  • Melting Pt:24…31 °C
  • Flash Pt:>110 °C
  • Storage Temperature:Ambient
  • EINECS:226-984-3
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications