Order Entry
Export
Orders LinkContactUsLinkComponent
Dimetridazole 98%

Dimetridazole 98%

Bottle, 500 g
Catalog # APOSOR927600-500G
Supplier:   Apollo Scientific
Dimetridazole 98%
Catalog # APOSOR927600-500G
Supplier:  Apollo Scientific
CAS Number:  551-92-8
Dimetridazole 98%

Bottle, 500 g

CAS Number:  551-92-8
Price
86,90 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  1,2-Dimethyl-5-nitroimidazole
  • CAS Number:551-92-8
  • Formula:C₅H₇N₃O₂
  • MDL Number:MFCD00047046
  • Molecular Weight:141.13 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g

Specifications