Order Entry
Export
Orders LinkContactUsLinkComponent
 

2,2-Diphenylethanol 98%

Bottle, 1 g
Catalog # APOSOR938457-1G
Supplier:   Apollo Scientific
2,2-Diphenylethanol 98%
Catalog # APOSOR938457-1G
Supplier:  Apollo Scientific
CAS Number:  1883-32-5
2,2-Diphenylethanol 98%

Bottle, 1 g

CAS Number:  1883-32-5
Price
36,20 €
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:1883-32-5
  • Formula:C₁₄H₁₄O
  • MDL Number:MFCD00004729
  • Molecular Weight:198.27 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications