Order Entry
Export
Orders LinkContactUsLinkComponent
2-Phenylethanol 95%

2-Phenylethanol 95%

Bottle, 100 g
Catalog # APOSOR921810-100G
Supplier:   Apollo Scientific
2-Phenylethanol 95%
Catalog # APOSOR921810-100G
Supplier:  Apollo Scientific
CAS Number:  60-12-8
2-Phenylethanol 95%

Bottle, 100 g

CAS Number:  60-12-8
Price
24,60 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Phenethyl alcohol, Benzylcarbinol, β-Hydroxyethylbenzene, Benzyl carbinol, Benzenethanol
  • CAS Number:60-12-8
  • Formula:C₆H₅CH₂CH₂OH
  • MDL Number:MFCD00002886
  • Molecular Weight:122.17 g/mol
  • Boiling Pt:219…221 °C
  • Melting Pt:–27 °C
  • Density:1.02 g/cm³ (25 °C)
  • Flash Pt:101–102 °C
  • Storage Temperature:Ambient
  • UN:2810
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications