Order Entry
Export
Orders LinkContactUsLinkComponent
1,3-Diethoxybenzene 97%

1,3-Diethoxybenzene 97%

Bottle, 5 g
Catalog # APOSOR920967-5G
Supplier:   Apollo Scientific
1,3-Diethoxybenzene 97%
Catalog # APOSOR920967-5G
Supplier:  Apollo Scientific
CAS Number:  2049-73-2
1,3-Diethoxybenzene 97%

Bottle, 5 g

CAS Number:  2049-73-2
Price
21,80 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:2049-73-2
  • Formula:C₁₀H₁₄O₂
  • MDL Number:MFCD00015145
  • Molecular Weight:166.22 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications