0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
1,3-Diethylurea
1,3-Diethylurea 97%
1,3-Diethylurea 97%
1,3-Diethylurea 97%
Bottle, 100 g
APOSOR323264-100G
:
Apollo Scientific
1,3-Diethylurea 97%
APOSOR323264-100G
:
Apollo Scientific
:
623-76-7
Expand image
1,3-Diethylurea 97%
Bottle, 100 g
Add to cart
:
623-76-7
84,00 €
Unit of Measure
1 * 100 g
-
+
Add to cart
:
N,N'-Diethylurea
:
623-76-7
:
C₅H₁₂N₂O
:
MFCD00009028
:
116.16 g/mol
:
109…111 °C
:
Ambient
Pack type:
Bottle
Pk:
100 g