Order Entry
Export
Orders LinkContactUsLinkComponent
 

1,6-Hexanediol 97%

Bottle, 250 g
Catalog # APOSOR322268-250G
Irritant
Supplier:   Apollo Scientific
1,6-Hexanediol 97%
Catalog # APOSOR322268-250G
Supplier:  Apollo Scientific
CAS Number:  629-11-8
1,6-Hexanediol 97%

Bottle, 250 g

CAS Number:  629-11-8
Price
44,90 €
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  Hexamethylene glycol, 1,6-Dihydroxy hexane
  • CAS Number:629-11-8
  • Formula:C₆H₁₄O₂
  • MDL Number:MFCD00002985
  • Molecular Weight:118.18 g/mol
  • Boiling Pt:248…250 °C
  • Melting Pt:40…43 °C
  • Density:1.12 g/cm³ (25 °C)
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 g

Specifications