Order Entry
Export
Orders LinkContactUsLinkComponent
 

2,6-Dibromoisonicotinic acid

Bottle, 5 g
Catalog # APOSOR2261-5G
Irritant
Supplier:   Apollo Scientific
2,6-Dibromoisonicotinic acid
Catalog # APOSOR2261-5G
Supplier:  Apollo Scientific
CAS Number:  2016-99-1
2,6-Dibromoisonicotinic acid

Bottle, 5 g

CAS Number:  2016-99-1
Price
53,60 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  2,6-Dibromopyridine-4-carboxylic acid
  • CAS Number:2016-99-1
  • Formula:C₆H₃Br₂NO₂
  • MDL Number:MFCD09743987
  • Molecular Weight:280.9000 g/mol
  • Storage Temperature:Refrigerator
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications