Order Entry
Export
Orders LinkContactUsLinkComponent
1,3-Diethynylbenzene 97%

1,3-Diethynylbenzene 97%

Bottle, 5 g
Catalog # APOSOR31042-5G
Irritant
Supplier:   Apollo Scientific
1,3-Diethynylbenzene 97%
Catalog # APOSOR31042-5G
Supplier:  Apollo Scientific
CAS Number:  1785-61-1
1,3-Diethynylbenzene 97%

Bottle, 5 g

CAS Number:  1785-61-1
Price
84,00 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:1785-61-1
  • Formula:C₁₀H₆
  • MDL Number:MFCD00041686
  • Molecular Weight:126.16 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications