0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
1,3-Diethynylbenzene
1,3-Diethynylbenzene 97%
1,3-Diethynylbenzene 97%
Bottle, 1 g
APOSOR31042-1G
:
Apollo Scientific
1,3-Diethynylbenzene 97%
APOSOR31042-1G
:
Apollo Scientific
:
1785-61-1
1,3-Diethynylbenzene 97%
Bottle, 1 g
:
1785-61-1
23,20 €
1 * 1 g
-
+
:
1785-61-1
:
C₁₀H₆
:
MFCD00041686
:
126.16 g/mol
:
Ambient
Pack type:
Bottle
Pk:
1 g