Order Entry
Export
Orders LinkContactUsLinkComponent
2-Methoxy-4-piperidinopyrimidine 98%

2-Methoxy-4-piperidinopyrimidine 98%

Bottle, 5 g
Catalog # APOSOR909498-5G
Supplier:   Apollo Scientific
2-Methoxy-4-piperidinopyrimidine 98%
Catalog # APOSOR909498-5G
Supplier:  Apollo Scientific
CAS Number:  1381944-24-6
2-Methoxy-4-piperidinopyrimidine 98%

Bottle, 5 g

CAS Number:  1381944-24-6
Price
435,00 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:1381944-24-6
  • Formula:C₁₀H₁₅N₃O
  • MDL Number:MFCD22375033
  • Molecular Weight:193.246 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications