Order Entry
Export
Orders LinkContactUsLinkComponent
2,3-Dichloropropionamide 95%

2,3-Dichloropropionamide 95%

Bottle, 5 g
Catalog # APOSOR938050-5G
Supplier:   Apollo Scientific
2,3-Dichloropropionamide 95%
Catalog # APOSOR938050-5G
Supplier:  Apollo Scientific
CAS Number:  19433-84-2
2,3-Dichloropropionamide 95%

Bottle, 5 g

CAS Number:  19433-84-2
Price
435,00 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:19433-84-2
  • Formula:C₃H₅Cl₂NO
  • MDL Number:MFCD00040311
  • Molecular Weight:141.98 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications