Order Entry
Export
Orders LinkContactUsLinkComponent
1,3-Adamantanediol

1,3-Adamantanediol

Bottle, 100 g
Catalog # APOSOR26342-100G
Irritant
Supplier:   Apollo Scientific
1,3-Adamantanediol
Catalog # APOSOR26342-100G
Supplier:  Apollo Scientific
CAS Number:  5001-18-3
1,3-Adamantanediol

Bottle, 100 g

CAS Number:  5001-18-3
Price
122,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  1,3-Dihydroxyadamantane
  • CAS Number:5001-18-3
  • Formula:C₁₀H₁₆O₂
  • MDL Number:MFCD00154013
  • Molecular Weight:168.24 g/mol
  • Melting Pt:260 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications