Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
2,4-Dichlorophenylhydrazine
2,4-Dichlorophenylhydrazine
2,4-Dichlorophenylhydrazine
Bottle, 5 g
APOSOR2419-5G
:
Apollo Scientific
2,4-Dichlorophenylhydrazine
APOSOR2419-5G
:
Apollo Scientific
:
13123-92-7
2,4-Dichlorophenylhydrazine
Bottle, 5 g
:
13123-92-7
427,00 €
1 * 5 g
-
+
:
(2,4-Dichlorophenyl)hydrazine
:
13123-92-7
:
C₆H₆Cl₂N₂
:
MFCD00044514
:
177.031 g/mol
:
91…94 °C
:
Refrigerator
Pack type:
Bottle
Pk:
5 g