Order Entry
Export
Orders LinkContactUsLinkComponent
 

1-Nitronaphthalene 99%

Bottle, 500 g
Catalog # APOSOR1884-500G
Flammable
Toxic
Environment
Supplier:   Apollo Scientific
1-Nitronaphthalene 99%
Catalog # APOSOR1884-500G
Supplier:  Apollo Scientific
CAS Number:  86-57-7
1-Nitronaphthalene 99%

Bottle, 500 g

CAS Number:  86-57-7
Price
89,80 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  α-Nitronaphthalene
  • CAS Number:86-57-7
  • Formula:C₁₀H₇NO₂
  • MDL Number:MFCD00003913
  • Molecular Weight:173.17 g/mol
  • Boiling Pt:303…305 °C
  • Melting Pt:53…58 °C
  • Density:1.223 g/cm³ (25 °C)
  • Flash Pt:164 °C
  • Storage Temperature:Ambient
  • EINECS:201-684-5
  • UN:2538
  • ADR:4.1,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g

Specifications