Order Entry
Export
Orders LinkContactUsLinkComponent
1,3-Cyclohexanediol 97%

1,3-Cyclohexanediol 97%

Bottle, 5 g
Catalog # APOSOR17557-5G
Irritant
Supplier:   Apollo Scientific
1,3-Cyclohexanediol 97%
Catalog # APOSOR17557-5G
Supplier:  Apollo Scientific
CAS Number:  504-01-8
1,3-Cyclohexanediol 97%

Bottle, 5 g

CAS Number:  504-01-8
Price
33,30 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  Cyclohexane-1,3-diol
  • CAS Number:504-01-8
  • Formula:C₆H₁₂O₂
  • MDL Number:MFCD00039458
  • Molecular Weight:116.16 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications