Order Entry
Export
Orders LinkContactUsLinkComponent
2,4-Dimethoxythiophenol 95%

2,4-Dimethoxythiophenol 95%

Bottle, 5 g
Catalog # APOSOR913049-5G
Supplier:   Apollo Scientific
2,4-Dimethoxythiophenol 95%
Catalog # APOSOR913049-5G
Supplier:  Apollo Scientific
CAS Number:  18906-37-1
2,4-Dimethoxythiophenol 95%

Bottle, 5 g

CAS Number:  18906-37-1
Price
536,00 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:18906-37-1
  • Formula:C₈H₁₀O₂S
  • MDL Number:MFCD04973436
  • Molecular Weight:170.2288 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications