1,4-Phenylenediphosphonic acid
Bottle, 250 mg
Catalog # APOSOR10672-250MG
Supplier: Apollo Scientific
1,4-Phenylenediphosphonic acid
Catalog # APOSOR10672-250MG
Supplier: Apollo Scientific
CAS Number: 880-68-2
Jump to:
Product Details & Documents
Synonyms: 1,4-Benzenediphosphonic acid
- CAS Number:880-68-2
- Formula:C₆H₈O₆P₂
- MDL Number:MFCD00475582
- Molecular Weight:238.07 g/mol
- Melting Pt:>260 °C
- Melting Pt:>260 °C
- Storage Temperature:Ambient
- UN:3261
- ADR:8,III
Specifications
Specifications
- Pack type:Bottle
- Pk:250 mg



