3,3'-Dihydroxybenzidine 97%
Bottle, 5 g
Catalog # APOSOR912142-5G
Supplier: Apollo Scientific
3,3'-Dihydroxybenzidine 97%
Catalog # APOSOR912142-5G
Supplier: Apollo Scientific
CAS Number: 2373-98-0
Jump to:
Product Details & Documents
Synonyms: 4,4'-Diamino-3,3'-dihydroxybiphenyl, 3,3'-Dihydroxy-4,4'-diaminobiphenyl, 4,4'-Diamino-[1,1'-biphenyl]-3,3'-diol, Benzidine-3,3'-diol
- CAS Number:2373-98-0
- Formula:C₁₂H₁₂N₂O₂
- MDL Number:MFCD00039149
- Molecular Weight:216.24 g/mol
- Melting Pt:292 °C
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:5 g



