Order Entry
Export
Orders LinkContactUsLinkComponent
3,3'-Dihydroxybenzidine 97%

3,3'-Dihydroxybenzidine 97%

Bottle, 10 g
Catalog # APOSOR912142-10G
Irritant
Health
Supplier:   Apollo Scientific
3,3'-Dihydroxybenzidine 97%
Catalog # APOSOR912142-10G
Supplier:  Apollo Scientific
CAS Number:  2373-98-0
3,3'-Dihydroxybenzidine 97%

Bottle, 10 g

CAS Number:  2373-98-0
Price
78,20 €
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  4,4'-Diamino-3,3'-dihydroxybiphenyl, 3,3'-Dihydroxy-4,4'-diaminobiphenyl, 4,4'-Diamino-[1,1'-biphenyl]-3,3'-diol, Benzidine-3,3'-diol
  • CAS Number:2373-98-0
  • Formula:C₁₂H₁₂N₂O₂
  • MDL Number:MFCD00039149
  • Molecular Weight:216.24 g/mol
  • Melting Pt:292 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    10 g

Specifications