0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
2,3-Dimethylanisole
2,3-Dimethylanisole 95%
2,3-Dimethylanisole 95%
Bottle, 25 g
APOSOR911822-25G
:
Apollo Scientific
2,3-Dimethylanisole 95%
APOSOR911822-25G
:
Apollo Scientific
:
2944-49-2
2,3-Dimethylanisole 95%
Bottle, 25 g
:
2944-49-2
50,70 €
1 * 25 g
-
+
:
1-Methoxy-2,3-dimethylbenzene
:
2944-49-2
:
C₉H₁₂O
:
MFCD00008376
:
136.19 g/mol
:
Ambient
Pack type:
Bottle
Pk:
25 g