Order Entry
Export
Orders LinkContactUsLinkComponent
2,3-Dimethylanisole 95%

2,3-Dimethylanisole 95%

Bottle, 100 g
Catalog # APOSOR911822-100G
Supplier:   Apollo Scientific
2,3-Dimethylanisole 95%
Catalog # APOSOR911822-100G
Supplier:  Apollo Scientific
CAS Number:  2944-49-2
2,3-Dimethylanisole 95%

Bottle, 100 g

CAS Number:  2944-49-2
Price
158,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  1-Methoxy-2,3-dimethylbenzene
  • CAS Number:2944-49-2
  • Formula:C₉H₁₂O
  • MDL Number:MFCD00008376
  • Molecular Weight:136.19 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications