Order Entry
Export
Orders LinkContactUsLinkComponent
(R)-(+)-2-Hydroxy-3-phenylpropionic acid 98%

(R)-(+)-2-Hydroxy-3-phenylpropionic acid 98%

Bottle, 100 g
Catalog # APOSOR911731-100G
Supplier:   Apollo Scientific
(R)-(+)-2-Hydroxy-3-phenylpropionic acid 98%
Catalog # APOSOR911731-100G
Supplier:  Apollo Scientific
CAS Number:  7326-19-4
(R)-(+)-2-Hydroxy-3-phenylpropionic acid 98%

Bottle, 100 g

CAS Number:  7326-19-4
Price
138,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  D-(+)-3-Phenyllactic acid, (R)-2-Hydroxy-3-phenylpropanoic acid, D(+)-Phenyllactic acid, D-​(+)​-​Phenyllactic acid
  • CAS Number:7326-19-4
  • Formula:C₉H₁₀O₃
  • MDL Number:MFCD00078062
  • Molecular Weight:166.1739 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications