Order Entry
Export
Orders LinkContactUsLinkComponent
2,4-Dichloropyridine

2,4-Dichloropyridine

Bottle, 500 g
Catalog # APOSOR6843-500G
Irritant
Supplier:   Apollo Scientific
2,4-Dichloropyridine
Catalog # APOSOR6843-500G
Supplier:  Apollo Scientific
CAS Number:  26452-80-2
2,4-Dichloropyridine

Bottle, 500 g

CAS Number:  26452-80-2
Price
235,00 €
Log in for availability
1 * 500 g
Product Details & Documents
  • CAS Number:26452-80-2
  • Formula:C₅H₃Cl₂N
  • MDL Number:MFCD00955618
  • Molecular Weight:147.99 g/mol
  • Boiling Pt:189…190 °C
  • Melting Pt:–1 °C
  • Density:1.37 g/cm³ (25 °C)
  • Flash Pt:89 °C
  • Storage Temperature:Ambient
  • EINECS:247-717-7
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g

Specifications