Poly(vinylidene fluoride)
Bottle, 25 g
Catalog # APOSPC37872-25G
Supplier: Apollo Scientific
Poly(vinylidene fluoride)
Catalog # APOSPC37872-25G
Supplier: Apollo Scientific
CAS Number: 24937-79-9
Jump to:
Product Details & Documents
- CAS Number:24937-79-9
- Formula:(-CH₂CF₂-)n
- MDL Number:MFCD00084470
- Melting Pt:155…165 °C
- Density:1.74 g/cm³ (25 °C)
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:25 g



