Order Entry
Export
Orders LinkContactUsLinkComponent
1,7-Dihydroxynaphthalene 95%

1,7-Dihydroxynaphthalene 95%

Bottle, 100 g
Catalog # APOSOR906184-100G
Supplier:   Apollo Scientific
1,7-Dihydroxynaphthalene 95%
Catalog # APOSOR906184-100G
Supplier:  Apollo Scientific
CAS Number:  575-38-2
1,7-Dihydroxynaphthalene 95%

Bottle, 100 g

CAS Number:  575-38-2
Price
375,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Naphthalene-1,7-diol
  • CAS Number:575-38-2
  • Formula:C₁₀H₈O₂
  • MDL Number:MFCD00035720
  • Molecular Weight:160.17 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications