Order Entry
Export
Orders LinkContactUsLinkComponent
 

(2-Bromophenoxy)acetic acid 97%

Bottle, 25 g
Catalog # APOSOR900608-25G
Supplier:   Apollo Scientific
(2-Bromophenoxy)acetic acid 97%
Catalog # APOSOR900608-25G
Supplier:  Apollo Scientific
CAS Number:  1879-56-7
(2-Bromophenoxy)acetic acid 97%

Bottle, 25 g

CAS Number:  1879-56-7
Price
207,00 €
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:1879-56-7
  • Formula:(C₆H₄Br)OCH₂COOH
  • MDL Number:MFCD00234407
  • Molecular Weight:231.05 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications