Order Entry
Export
Orders LinkContactUsLinkComponent
3,6-Dicyanopicoline 97%

3,6-Dicyanopicoline 97%

Bottle, 1 g
Catalog # APOSOR959412-1G
Supplier:   Apollo Scientific
3,6-Dicyanopicoline 97%
Catalog # APOSOR959412-1G
Supplier:  Apollo Scientific
CAS Number:  1352318-05-8
3,6-Dicyanopicoline 97%

Bottle, 1 g

CAS Number:  1352318-05-8
Price
362,00 €
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:1352318-05-8
  • Formula:C₈H₅N₃
  • MDL Number:MFCD20529525
  • Molecular Weight:143.145 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications