Order Entry
Export
Orders LinkContactUsLinkComponent
5-Hydroxy-2-nitropyridine 95%

5-Hydroxy-2-nitropyridine 95%

Bottle, 1 g
Catalog # APOSOR959407-1G
Supplier:   Apollo Scientific
5-Hydroxy-2-nitropyridine 95%
Catalog # APOSOR959407-1G
Supplier:  Apollo Scientific
CAS Number:  15206-26-5
5-Hydroxy-2-nitropyridine 95%

Bottle, 1 g

CAS Number:  15206-26-5
Price
102,00 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  6-nitropyridin-3-ol
  • CAS Number:15206-26-5
  • Formula:C5H4N2O3
  • MDL Number:MFCD03032441
  • Molecular Weight:140.0969 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications