Order Entry
Export
Orders LinkContactUsLinkComponent
 

Benzhydrylamine

Bottle, 1 g
Catalog # APOSOR8244-1G
Corrosive
Irritant
Supplier:   Apollo Scientific
Benzhydrylamine
Catalog # APOSOR8244-1G
Supplier:  Apollo Scientific
CAS Number:  91-00-9
Benzhydrylamine

Bottle, 1 g

CAS Number:  91-00-9
Price
21,80 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  α-Phenylbenzylamine, α-Aminodiphenylmethane, Diphenylmethanamine, Aminodiphenylmethane, 1,1-Diphenylmethanamine, Benzhydrylamin
  • CAS Number:91-00-9
  • Formula:C₁₃H₁₃N
  • MDL Number:MFCD00008059
  • Molecular Weight:183.25 g/mol
  • Boiling Pt:294…296 °C
  • Melting Pt:41579 °C
  • Density:1.063 g/cm³ (25 °C)
  • Flash Pt:>110 °C
  • Storage Temperature:Ambient
  • EINECS:202-032-2
  • UN:2735
  • ADR:8,II
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications