6-Bromo-4-methoxy-pyrazolo[1,5-a]pyridine
Bottle, 250 mg
Catalog # APOSOR49065-250MG
Supplier: Apollo Scientific
6-Bromo-4-methoxy-pyrazolo[1,5-a]pyridine
Catalog # APOSOR49065-250MG
Supplier: Apollo Scientific
CAS Number: 1207557-36-5
Jump to:
Product Details & Documents
Synonyms: 6-Bromo-4-methoxypyrazolo[1,5-a]pyridine
- CAS Number:1207557-36-5
- Formula:C₈H₇BrN₂O
- MDL Number:MFCD17926335
- Molecular Weight:227.058 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:250 mg



