Order Entry
Export
Orders LinkContactUsLinkComponent
2-Methyl-1,8-naphthyridine 98%

2-Methyl-1,8-naphthyridine 98%

Bottle, 1 g
Catalog # APOSOR951676-1G
Supplier:   Apollo Scientific
2-Methyl-1,8-naphthyridine 98%
Catalog # APOSOR951676-1G
Supplier:  Apollo Scientific
CAS Number:  1569-16-0
2-Methyl-1,8-naphthyridine 98%

Bottle, 1 g

CAS Number:  1569-16-0
Price
24,60 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  2-Methyl-1,8-diazanaphthalene, 2-Methylpyrido[2,3-b]pyridine, 2-Methyl[1,8]-Naphthyridine
  • CAS Number:1569-16-0
  • Formula:C₉H₈N₂
  • MDL Number:MFCD04066717
  • Molecular Weight:144.18 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications