Quinoline 98%
Bottle, 250 g
Catalog # APOSOR45105-250G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
- CAS Number:91-22-5
- Formula:C₉H₇N
- MDL Number:MFCD00006736
- Molecular Weight:129.16 g/mol
- Boiling Pt:236…238 °C
- Melting Pt:–17…–13 °C
- Melting Pt:–17 to –13 °C
- Density:1.093 g/cm³ (25 °C)
- Flash Pt:101 °C
- Storage Temperature:Ambient
- EINECS:202-051-6
- UN:2656
- ADR:6.1,III
Specifications
Specifications
- Pack type:Bottle
- Pk:250 g



