Order Entry
Export
Orders LinkContactUsLinkComponent
Perchloric acid ≥70%

Perchloric acid ≥70%

Glass bottle, 50 mL
Catalog # SIAL311421-50ML
Oxidizing
Corrosive
Supplier:   Merck
Perchloric acid ≥70%
Catalog # SIAL311421-50ML
Supplier:  Merck
CAS Number:  7601-90-3
Perchloric acid ≥70%

Glass bottle, 50 mL

CAS Number:  7601-90-3
Price
95,00 €
Log in for availability
1 * 50 mL
Product Details & Documents
  • CAS Number:7601-90-3
  • Formula:HClO₄
  • MDL Number:MFCD00011325
  • Molecular Weight:100.46 g/mol
  • Boiling Pt:160…181 °C (1013 hPa)
  • Melting Pt:–112.2 °C
  • Density:1.53…1.768 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-512-4
  • UN:1873
  • ADR:5.1,I

Documents

Specifications

Specification Test Results

  • Assay:
    70%

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 mL

Specifications