Order Entry
Export
Orders LinkContactUsLinkComponent
 

Human recombinant Alpha-Synuclein (from E. coli)

200 µG
  ACRBALN-H52H8200UG
Human recombinant Alpha-Synuclein (from<i> E. coli</i>)
  ACRBALN-H52H8200UG
 :  
Human recombinant Alpha-Synuclein (from E. coli)

200 µG

 :  
 
165,00 €
   
1 * 200 µG
 

Human Alpha-Synuclein protein, His Tag.

 

 

  • Pk:
    200 µG
  • Protein/peptide name:
    Alpha-Synuclein
  • Protein synonyms:
    PARK1|SNCA|alpha-Synuclein|NACP
  • Purity:
    >90% as determined by SDS-PAGE.
  • Endotoxin level:
    Low
  • Source:
    E. coli
  • Molecular weight:
    16,3 kDa
  • Shipping temperature:
    RT
  • Cat. No.:
    ACRBALN-H52H8200UG
  • Protein/peptide type:
    Recombinant
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Storage conditions:
    –20 °C
  • Gene ID:
    P37840-1
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in 100 mM NaAC, pH 7,0. Normally trehalose is added as protectant before Lyophilisation.
  • Conjugation:
    Unconjugated
  • Tag sequence:
    His Tag
  • Species:
    Human