Human recombinant Alpha-Synuclein (from E. coli)
1 mg
ACRBALN-H52H8-1MG
Human Alpha-Synuclein protein, His Tag.
- Pk:1 mg
- Protein/peptide name:Alpha-Synuclein
- Protein synonyms:PARK1|SNCA|alpha-Synuclein|NACP
- Purity:>90% as determined by SDS-PAGE.
- Endotoxin level:Low
- Source:E. coli
- Molecular weight:16,3 kDa
- Shipping temperature:RT
- Cat. No.:ACRBALN-H52H8-1MG
- Protein/peptide type:Recombinant
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Storage conditions:–20 °C
- Gene ID:P37840-1
- Formulation:Lyophilised from 0,22 μm filtered solution in 100 mM NaAC, pH 7,0. Normally trehalose is added as protectant before Lyophilisation.
- Conjugation:Unconjugated
- Tag sequence:His Tag
- Species:Human



