ContactUsLinkComponent
SARS-CoV-2 NSP16NSP10 Heterodimer Protein, His TagTwin Strep Tag NS0-C51W3 is expressed from E.coli cells. It contains AA Ser 1 - As n 298 NSP16 Ala 1 - Gln 139 NSP10 Accession YP_009725311.1 NSP16 YP_009725306.1 NSP10. 1 * 100 µG
Catalog # ACRBNS0-C51W3100UG
Supplier:  ACROBIOSYSTEMS
SARS-CoV-2 NSP16NSP10 Heterodimer Protein, His TagTwin Strep Tag NS0-C51W3 is expressed from E.coli cells. It contains AA Ser 1 - As n 298 NSP16 Ala 1 - Gln 139 NSP10 Accession YP_009725311.1 NSP16 YP_009725306.1 NSP10. 1 * 100 µG
Catalog # ACRBNS0-C51W3100UG
Supplier:  ACROBIOSYSTEMS