ContactUsLinkComponent
SARS-CoV-2 S1 protein, His Tag S1N-C52H2 is expressed from human 293 cells HEK293. It contains AA Val 16 - Arg 685 Accession QHD434 16.1. 1 * 100 mg
Catalog # ACRBS1N-C52H2100MG
Supplier:  ACRO Biosystems
SARS-CoV-2 S1 protein, His Tag S1N-C52H2 is expressed from human 293 cells HEK293. It contains AA Val 16 - Arg 685 Accession QHD434 16.1. 1 * 100 mg
Catalog # ACRBS1N-C52H2100MG
Supplier:  ACRO Biosystems