ContactUsLinkComponent
SARS-CoV-2 S protein RBD, Mouse IgG2a Fc Tag SPD-C5251 is expressed from human 293 cells HEK293. It contains AA Arg 319 - Phe 541 Ac cession QHD43416.1. 1 * 200 µG
Catalog # ACRBSPD-C5251200UG
Supplier:  ACRO Biosystems
SARS-CoV-2 S protein RBD, Mouse IgG2a Fc Tag SPD-C5251 is expressed from human 293 cells HEK293. It contains AA Arg 319 - Phe 541 Ac cession QHD43416.1. 1 * 200 µG
Catalog # ACRBSPD-C5251200UG
Supplier:  ACRO Biosystems