ContactUsLinkComponent
SARS-CoV-2 S protein RBD V483A, His Tag SPD-C52H5 is expressed from human 293 cells HEK293. It contains AA Arg 319 - Phe 541 Accessi on QHD43416.1 V483A. 1 * 100 µG
Catalog # ACRBSPD-C52H5100UG
Supplier:  ACRO Biosystems
SARS-CoV-2 S protein RBD V483A, His Tag SPD-C52H5 is expressed from human 293 cells HEK293. It contains AA Arg 319 - Phe 541 Accessi on QHD43416.1 V483A. 1 * 100 µG
Catalog # ACRBSPD-C52H5100UG
Supplier:  ACRO Biosystems