Mouse recombinant IL-33 (from HEK293 cells)
50 µG
Catalog # ACRBIL3-M5245-50UG
Supplier: ACRO Biosystems
Mouse recombinant IL-33 (from HEK293 cells)
Catalog # ACRBIL3-M5245-50UG
Supplier: ACRO Biosystems
CAS Number:
Jump to:
Product Details & Documents
Mouse IL-33 Protein, His Tag (MALS verified), ACROBiosystems
Specifications
Specifications
- Pk:50 µG
- Protein/peptide name:IL-33
- Protein synonyms:DV27|NFEHEV|IL33|IL1F11|DKFZp586H0523|DVS27|NFHEV|RP11-575C20.2|C9ORF26
- Purity:>90% as determined by SDS-PAGE
- Endotoxin level:Low
- Source:HEK293 cells
- Molecular weight:19,5 kDa
- Shipping temperature:RT
- Cat. No.:ACRBIL3-M5245-50UG
- Protein/peptide type:Recombinant
- Endotoxin content:Less than 1,0 EU per μg by the LAL method
- Storage conditions:–20 °C
- Gene ID:Q8BVZ5-1
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4 . Normally trehalose is added as protectant before lyophilisation
- Conjugation:Unconjugated
- Tag sequence:His Tag (MALS verified)
- Species:Mouse



