ContactUsLinkComponent
Biotinylated Mouse CD3ECD3G Heterodimer Protein, His,AvitagFlag Tag CDG-M58D2 is expressed from human 293 cells HEK293. It contains AA Asp 23 - Asp 108 CD3E Gln 23 - Ser 116 CD3G Accession P22646-1 CD3E P11942-1 CD3G. 1 * 200 µG
Catalog # ACRBCDG-M58D2200UG
Supplier:  ACROBIOSYSTEMS
Biotinylated Mouse CD3ECD3G Heterodimer Protein, His,AvitagFlag Tag CDG-M58D2 is expressed from human 293 cells HEK293. It contains AA Asp 23 - Asp 108 CD3E Gln 23 - Ser 116 CD3G Accession P22646-1 CD3E P11942-1 CD3G. 1 * 200 µG
Catalog # ACRBCDG-M58D2200UG
Supplier:  ACROBIOSYSTEMS