Order Entry
Export
Orders LinkContactUsLinkComponent
 

VWR®, Pipette Tips, Universal

50 - 1000 µl, 70 mm, Blue, Bulk (1×500)
  612-6786
VWR®, Pipette Tips, Universal
  612-6786
VWR®, Pipette Tips, Universal
VWR®, Pipette Tips, Universal

50 - 1000 µl, 70 mm, Blue, Bulk (1×500)

 
11,60 €
   
1 * 500 items
 

PP, autoclavable.

  • Manufactured without lubricants
  • Cadmium-free pigments
  • Environmentally friendly packaging system

Colour: Yellow (2 - 200 µl), blue (50 - 1000 µl)

Length: 50 mm (2 - 200 µl), 70 mm (50 - 1000 µl)

 
  • Packed:
    Bulk (1×500)
  • Volume:
    50 - 1000 µl
  • Length:
    70 mm
  • Tip colour:
    Blue
Accessories

 

  •  
    Description
    For use with
     
     
  • 613-2288
    Tip-Box N, PP, with push on lid, stackable and autoclavable, for 1000 µl tips
    50 - 1000 µl tips
       
    1 * 1 items - 15,50 €

 

  •  
    Description
    For use with
     
     
  • 613-2287
    Tip-Box N, PP, with push on lid, stackable and autoclavable, for 200 µl tips
    2 - 200 µl tips
       
    1 * 1 items - 13,10 €