0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
MP SILICA DCC MEAN PORE SIZE 60 1 * 3 kg
0204526.
:
MP Biomedicals
MP SILICA DCC MEAN PORE SIZE 60 1 * 3 kg
0204526.
:
MP Biomedicals