Accessories for Rebel microscopes, Echo
Polarizer kit for Rebel LWD condenser with rotatable polarizer and fixed analyzer
Catalog # ECHLZND0003
Supplier: DISCOVER ECHO
Jump to:
Product Details & Documents
Documents
ECHO Rebel Microscope Specification
Product Literature
Specifications
- Description:Polarizer kit for Rebel LWD condenser with rotatable polarizer and fixed analyzer